Merck
  • Spiralisones A-D: acylphloroglucinol hemiketals from an Australian marine brown alga, Zonaria spiralis.
Spiralisones A-D: acylphloroglucinol hemiketals from an Australian marine brown alga, Zonaria spiralis.
Organic & biomolecular chemistry (2012-11-14)
Hua Zhang, Xue Xiao, Melissa M Conte, Zeinab Khalil, Robert J Capon
ABSTRACT
An intertidal sample of the Australian marine brown alga, Zonaria spiralis, exhibited promising kinase inhibitory and antibacterial activity. Chemical analysis returned six phloroglucinol-derived lipids, the new hemiketal spiralisones A-D (1-4) and the known chromones 5-6, and the known norsesquiterpenoid apo-9'-fucoxanthinone (7). Structures 1-7 were assigned on the basis of detailed spectroscopic analysis, biosynthetic considerations and total synthesis. Spiralisones undergo facile acid-mediated dehydration to yield the corresponding chromones, revealing for the first time that brown algal chromones may be handling artifacts rather than natural products. Hemiketals 1 and 2, and chromone 6, displayed inhibitory activity against the neurodegenerative disease kinase targets CDK5/p25, CK1δ and GSK3β, while hemiketals 1, 3 and 4, and chromone 6, displayed growth inhibitory activity against the Gram-positive bacteria Bacillus subtilis (ATCC 6051 and 6633). The promising kinase inhibitory and antibacterial properties of the Z. spiralis extract were attributed to the cumulative effect of many moderately potent phloroglucinol-derived lipid co-metabolites.
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
Sigma-Aldrich
Phloroglucinol, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Sigma-Aldrich
Phloroglucinol, Used to detect the presence of wood fiber.