Merck
  • FGF23 is a novel regulator of intracellular calcium and cardiac contractility in addition to cardiac hypertrophy.
FGF23 is a novel regulator of intracellular calcium and cardiac contractility in addition to cardiac hypertrophy.
American journal of physiology. Endocrinology and metabolism (2013-02-28)
Chad D Touchberry, Troy M Green, Vladimir Tchikrizov, Jaimee E Mannix, Tiffany F Mao, Brandon W Carney, Magdy Girgis, Robert J Vincent, Lori A Wetmore, Buddhadeb Dawn, Lynda F Bonewald, Jason R Stubbs, Michael J Wacker
ABSTRACT
Fibroblast growth factor 23 (FGF23) is a hormone released primarily by osteocytes that regulates phosphate and vitamin D metabolism. Recent observational studies in humans suggest that circulating FGF23 is independently associated with cardiac hypertrophy and increased mortality, but it is unknown whether FGF23 can directly alter cardiac function. We found that FGF23 significantly increased cardiomyocyte cell size in vitro, the expression of gene markers of cardiac hypertrophy, and total protein content of cardiac muscle. In addition, FGFR1 and FGFR3 mRNA were the most abundantly expressed FGF receptors in cardiomyocytes, and the coreceptor α-klotho was expressed at very low levels. We tested an animal model of chronic kidney disease (Col4a3(-/-) mice) that has elevated serum FGF23. We found elevations in common hypertrophy gene markers in Col4a3(-/-) hearts compared with wild type but did not observe changes in wall thickness or cell size by week 10. However, the Col4a3(-/-) hearts did show reduced fractional shortening (-17%) and ejection fraction (-11%). Acute exposure of primary cardiomyocytes to FGF23 resulted in elevated intracellular Ca(2+) ([Ca(2+)](i); F/F(o) + 86%) which was blocked by verapamil pretreatment. FGF23 also increased ventricular muscle strip contractility (67%), which was inhibited by FGF receptor antagonism. We hypothesize that although FGF23 can acutely increase [Ca(2+)](i), chronically this may lead to decreases in contractile function or stimulate cardiac hypertrophy, as observed with other stress hormones. In conclusion, FGF23 is a novel bone/heart endocrine factor and may be an important mediator of cardiac Ca(2+) regulation and contractile function during chronic kidney disease.
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4