Merck
  • A strand-specific burst in transcription of pericentric satellites is required for chromocenter formation and early mouse development.
A strand-specific burst in transcription of pericentric satellites is required for chromocenter formation and early mouse development.
Developmental cell (2010-10-19)
Aline V Probst, Ikuhiro Okamoto, Miguel Casanova, Fatima El Marjou, Patricia Le Baccon, Geneviève Almouzni
ABSTRACT
At the time of fertilization, the paternal genome lacks the typical configuration and marks characteristic of pericentric heterochromatin. It is thus essential to understand the dynamics of this region during early development, its importance during that time period and how a somatic configuration is attained. Here, we show that pericentric satellites undergo a transient peak in expression precisely at the time of chromocenter formation. This transcription is regulated in a strand-specific manner in time and space and is strongly biased by the parental asymmetry. The transcriptional upregulation follows a developmental clock, yet when replication is blocked chromocenter formation is impeded. Furthermore, interference with major satellite transcripts using locked nucleic acid (LNA)-DNA gapmers results in developmental arrest before completion of chromocenter formation. We conclude that the exquisite strand-specific expression dynamics at major satellites during the 2-cell stage, with both up and downregulation, are necessary events for proper chromocenter organization and developmental progression.
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
M16 Medium, With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
SSC Buffer, for Northern and Southern blotting, powder blend
Sigma-Aldrich
SSC Buffer 20× Concentrate, for Northern and Southern blotting, solution