Merck
  • Release behavior of tanshinone IIA sustained-release pellets based on crack formation theory.
Release behavior of tanshinone IIA sustained-release pellets based on crack formation theory.
Journal of pharmaceutical sciences (2012-05-23)
Pan Liu, Jin Li, Jianping Liu, Jikun Yang, Yongqing Fan
ABSTRACT
The objective of this study was to investigate the drug release mechanism and in vivo performance of Tanshinone IIA sustained-release pellets, coated with blends of polyvinyl acetate (PVAc) and poly(vinyl alcohol)-poly(ethylene glycol) (PVA-PEG) graft copolymer. A formulation screening study showed that pellets coated with PVAc-PVA-PEG at a ratio of 70:30 (w/w) succeeded in achieving a 24 h sustained release, irrespective of the coating weight (from 2% to 10%). Both the microscopic observation and mathematical model gave further insight into the underlying release mechanism, indicating that diffusion through water-filled cracks was dominant for the control of drug release. In vivo test showed that the maximum plasma concentration of sustained-release pellets was decreased from 82.13 ± 17.05 to 40.50 ± 11.72 ng mL as that of quick-release pellets. The time of maximum concentration, half time, and mean residence time were all prolonged from 3.80 ± 0.40 to 8.02 ± 0.81 h, 4.28 ± 1.21 to 8.18 ± 2.06 h, and 8.60 ± 1.59 to 17.50 ± 2.78 h, compared with uncoated preparations. A good in vitro-in vivo correlation was characterized by a high coefficient of determination (r = 0.9772). In conclusion, pellets coated with PVAc-PVA-PEG could achieve a satisfactory sustained-release behavior based on crack formation theory.
Sigma-Aldrich
Poly(vinyl acetate), average Mw ~500,000 by GPC
Sigma-Aldrich
Poly(vinyl acetate), average Mw ~100,000 by GPC, beads
Poly(vinyl acetate) dispersion 30 per cent, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Kollicoat® IR
Sigma-Aldrich
Kollicoat® SR 30 D, 28.5-31.5% solids basis