Merck
  • Genomic erosion and horizontal gene transfer shape functional differences of the ExlA toxin in Pseudomonas spp.
Genomic erosion and horizontal gene transfer shape functional differences of the ExlA toxin in Pseudomonas spp.
iScience (2022-07-06)
Viviana Job, Laura Gomez-Valero, Adèle Renier, Christophe Rusniok, Stephanie Bouillot, Viviane Chenal-Francisque, Erwan Gueguen, Annie Adrait, Mylène Robert-Genthon, Katy Jeannot, Peter Panchev, Sylvie Elsen, Marie-Odile Fauvarque, Yohann Couté, Carmen Buchrieser, Ina Attrée
ABSTRACT
Two-partner secretion (TPS) is widespread in the bacterial world. The pore-forming TPS toxin ExlA of Pseudomonas aeruginosa is conserved in pathogenic and environmental Pseudomonas. While P. chlororaphis and P. entomophila displayed ExlA-dependent killing, P. putida did not cause damage to eukaryotic cells. ExlA proteins interacted with epithelial cell membranes; however, only ExlA Pch induced the cleavage of the adhesive molecule E-cadherin. ExlA proteins participated in insecticidal activity toward the larvae of Galleria mellonella and the fly Drosophila melanogaster. Evolutionary analyses demonstrated that the differences in the C-terminal domains are partly due to horizontal movements of the operon within the genus Pseudomonas. Reconstruction of the evolutionary history revealed the complex horizontal acquisitions. Together, our results provide evidence that conserved TPS toxins in environmental Pseudomonas play a role in bacteria-insect interactions and discrete differences in CTDs may determine their specificity and mode of action toward eukaryotic cells.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Okadaic Acid, Prorocentrum sp., Okadaic Acid, CAS 78111-17-8, is a highly potent inhibitor of protein phosphatase 1 (IC₅₀ = 10-15 nM) and 2A (IC₅₀ = 0.1 nM).
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone D66, purified from hybridoma cell culture