Merck
  • A type II protein arginine methyltransferase regulates merozoite invasion in Plasmodium falciparum.
A type II protein arginine methyltransferase regulates merozoite invasion in Plasmodium falciparum.
Communications biology (2023-06-23)
Amuza Byaruhanga Lucky, Chengqi Wang, Min Liu, Xiaoying Liang, Hui Min, Qi Fan, Faiza Amber Siddiqui, Swamy Rakesh Adapa, Xiaolian Li, Rays H Y Jiang, Xiaoguang Chen, Liwang Cui, Jun Miao
ABSTRACT
Protein arginine methyltransferases (PRMTs) regulate many important cellular processes, such as transcription and RNA processing in model organisms but their functions in human malaria parasites are not elucidated. Here, we characterize PfPRMT5 in Plasmodium falciparum, which catalyzes symmetric dimethylation of histone H3 at R2 (H3R2me2s) and R8, and histone H4 at R3 in vitro. PfPRMT5 disruption results in asexual stage growth defects primarily due to lower invasion efficiency of the merozoites. Transcriptomic analysis reveals down-regulation of many transcripts related to invasion upon PfPRMT5 disruption, in agreement with H3R2me2s being an active chromatin mark. Genome-wide chromatin profiling detects extensive H3R2me2s marking of genes of different cellular processes, including invasion-related genes in wildtype parasites and PfPRMT5 disruption leads to the depletion of H3R2me2s. Interactome studies identify the association of PfPRMT5 with invasion-related transcriptional regulators such as AP2-I, BDP1, and GCN5. Furthermore, PfPRMT5 is associated with the RNA splicing machinery, and PfPRMT5 disruption caused substantial anomalies in RNA splicing events, including those for invasion-related genes. In summary, PfPRMT5 is critical for regulating parasite invasion and RNA splicing in this early-branching eukaryote.
Sigma-Aldrich
Anti-Histone H4 Antibody, pan, rabbit monoclonal, culture supernatant, clone 62-141-13, Upstate®
Sigma-Aldrich
Anti-dimethyl-Histone H4 (Arg3) Symmetric Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Histone H3 Antibody, 0.5 mg/mL, Upstate®