Merck
  • Transcription-Dependent Formation of Nuclear Granules Containing FUS and RNA Pol II.
Transcription-Dependent Formation of Nuclear Granules Containing FUS and RNA Pol II.
Biochemistry (2018-11-30)
Valery F Thompson, Rachel A Victor, Andres A Morera, Mahta Moinpour, Meilani N Liu, Conner C Kisiel, Kaitlyn Pickrel, Charis E Springhower, Jacob C Schwartz
ABSTRACT
Purified recombinant FUsed in Sarcoma (FUS) assembles into an oligomeric state in an RNA-dependent manner to form large condensates. FUS condensates bind and concentrate the C-terminal domain of RNA polymerase II (RNA Pol II). We asked whether a granule in cells contained FUS and RNA Pol II as suggested by the binding of FUS condensates to the polymerase. We developed cross-linking protocols to recover protein particles containing FUS from cells and separated them by size exclusion chromatography. We found a significant fraction of RNA Pol II in large granules containing FUS with diameters of >50 nm or twice that of the RNA Pol II holoenzyme. Inhibition of transcription prevented the polymerase from associating with the granules. Altogether, we found physical evidence of granules containing FUS and RNA Pol II in cells that possess properties comparable to those of in vitro FUS condensates.
Sigma-Aldrich
Anti-Histone H2B Antibody, clone 5HH2-2A8, ascites fluid, clone 5HH2-2A8, from mouse
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Anti-RNA polymerase II Antibody, clone CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
DAPI, for nucleic acid staining