Merck
Methods to Investigate EGFR Ubiquitination.
Methods in molecular biology (Clifton, N.J.) (2017-08-10)
Alexia Conte, Sara Sigismund
ABSTRACT
Ubiquitination of the epidermal growth factor receptor (EGFR) is an important intracellular signal that occurs upon EGF stimulation and controls EGFR trafficking at multiple steps, finally destining the receptor to lysosomal degradation. In this chapter, we give an overview of the biochemical methods to investigate EGFR ubiquitination.Firstly, we describe the in vitro ubiquitination assay, a method where, in the presence of the minimal ubiquitination machinery, the biological milieu for EGFR ubiquitination is reproduced in a test tube. In the second protocol, we explain how to immunoprecipitate the EGFR from total lysate and reveal its ubiquitinated form by western blot analysis. Then, with an ELISA-derived assay, we illustrate a robust and reliable method to assess EGFR ubiquitination from low amount of sample; lastly, we illustrate an immunofluorescence protocol to visualize ubiquitinated species (including the EGFR itself) within the EGFR-positive endocytic compartments upon EGF stimulation.
Sigma-Aldrich
Anti-EGFR Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-EGFR (Ab-1) Mouse mAb (528), lyophilized, clone 528, Calbiochem®
Sigma-Aldrich
Anti-Ubiquitin Antibody, Lys63-Specific, clone Apu3, rabbit monoclonal, clone Apu3, from rabbit
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse