Merck
  • Blockade of HIV-1 infection of New World monkey cells occurs primarily at the stage of virus entry.
Blockade of HIV-1 infection of New World monkey cells occurs primarily at the stage of virus entry.
The Journal of experimental medicine (2002-08-21)
Jason A LaBonte, Gregory J Babcock, Trushar Patel, Joseph Sodroski
ABSTRACT
HIV-1 naturally infects chimpanzees and humans, but does not infect Old World monkeys because of replication blocks that occur after virus entry into the cell. To understand the species-specific restrictions operating on HIV-1 infection, the ability of HIV-1 to infect the cells of New World monkeys was examined. Primary cells derived from common marmosets and squirrel monkeys support every phase of HIV-1 replication with the exception of virus entry. Efficient HIV-1 entry typically requires binding of the viral envelope glycoproteins and host cell receptors, CD4 and either CCR5 or CXCR4 chemokine receptors. HIV-1 did not detectably bind or utilize squirrel monkey CD4 for entry, and marmoset CD4 was also very inefficient compared with human CD4. A marmoset CD4 variant, in which residues 48 and 59 were altered to the amino acids found in human CD4, supported HIV-1 entry efficiently. The CXCR4 molecules of both marmosets and squirrel monkeys supported HIV-1 infection, but the CCR5 proteins of both species were only marginally functional. These results demonstrate that the CD4 and CCR5 proteins of New World monkeys represent the major restriction against HIV-1 replication in these primates. Directed adaptation of the HIV-1 envelope glycoproteins to common marmoset receptors might allow the development of New World monkey models of HIV-1 infection.
Sigma-Aldrich
Anti-CD4 antibody, Mouse monoclonal, clone Q4120, purified from hybridoma cell culture