Merck
  • BMP-4, TGF-β and Smad3 as Modulators of Synovial Fluid Cells Viability.
BMP-4, TGF-β and Smad3 as Modulators of Synovial Fluid Cells Viability.
Revista brasileira de ortopedia (2021-03-31)
Eduardo Branco de Sousa, Vivaldo Moura Neto, Diego Pinheiro Aguiar
ABSTRACT
Objective  Our goal was to evaluate the modulation of the synovial fluid cells (SFC) from patients with and without osteoarthritis (OA) by bone morphogenetic protein 4 (BMP-4), Smad-3 and transforming growth factor beta (TGF-β). Methods  Synovial fluid was collected from patients submitted to knee arthroscopy or replacement and were centrifuged to isolate cells from the fluid. Cells were cultured for 21 days and characterized as mesenchymal stem cells (MSCs) according to the criteria of the International Society of Cell Therapy. Then, we performed an [3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide] assay (MTT) assay after exposing cells with and without OA to TGF-β, Smad3 and BMP-4 pathway inhibitors and to different concentrations of BMP4. Results  Exposure to the TGF-β, Smad3 and BMP-4 inhibitors modifies the mitochondrial activity of the SFCs. The activity of the SFCs is modified by influences of increasing concentrations of BMP4, but there is no difference in cellular activity between patients with and without OA. Conclusion  TGF-β, Smad3 and BMP-4 modulate the activity of SFCs from patients with and without knee OA.
Sigma-Aldrich
BMP Inhibitor II, DMH1, The BMP Inhibitor II, DMH1, also referenced under CAS 1206711-16-1, controls the biological activity of BMP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
1,2,4,5-Tetramethylbenzene-d14, 98 atom % D
Sigma-Aldrich
Smad3 Inhibitor, SIS3, Smad3 Inhibitor, SIS3, CAS 1009104-85-1, is a cell-permeable, selective inhibitor of TGF-β1-dependent Smad3 phosphorylation and Smad3-mediated signaling. Does not affect Smad2, MAPK, ERK, or PI3-K.