Merck
  • A systematic approach to inserting split inteins for Boolean logic gate engineering and basal activity reduction.
A systematic approach to inserting split inteins for Boolean logic gate engineering and basal activity reduction.
Nature communications (2021-04-15)
Trevor Y H Ho, Alexander Shao, Zeyu Lu, Harri Savilahti, Filippo Menolascina, Lei Wang, Neil Dalchau, Baojun Wang
ABSTRACT
Split inteins are powerful tools for seamless ligation of synthetic split proteins. Yet, their use remains limited because the already intricate split site identification problem is often complicated by the requirement of extein junction sequences. To address this, we augment a mini-Mu transposon-based screening approach and devise the intein-assisted bisection mapping (IBM) method. IBM robustly reveals clusters of split sites on five proteins, converting them into AND or NAND logic gates. We further show that the use of inteins expands functional sequence space for splitting a protein. We also demonstrate the utility of our approach over rational inference of split sites from secondary structure alignment of homologous proteins, and that basal activities of highly active proteins can be mitigated by splitting them. Our work offers a generalizable and systematic route towards creating split protein-intein fusions for synthetic biology.
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
N-(β-Ketocaproyl)-L-homoserine lactone, ≥98%
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer