Merck
  • Bioluminescent optogenetic (BL-OG) activation of neurons during mouse postnatal brain development.
Bioluminescent optogenetic (BL-OG) activation of neurons during mouse postnatal brain development.
STAR protocols (2021-07-22)
Emmanuel L Crespo, Mansi Prakash, Andreas Bjorefeldt, William E Medendorp, Nathan C Shaner, Diane Lipscombe, Christopher I Moore, Ute Hochgeschwender
ABSTRACT
Bioluminescent optogenetics (BL-OG) allows activation of photosensory proteins, such as opsins, by either fiberoptics or by administering a luciferin. BL-OG thus confers both optogenetic and chemogenetic access within the same genetically targeted neuron. This bimodality offers a powerful approach for non-invasive chemogenetic manipulation of neural activity during brain development and adult behaviors with standard optogenetic spatiotemporal precision. We detail protocols for bioluminescent stimulation of neurons in postnatally developing brain and its validation through bioluminescence imaging and electrophysiological recording in mice. For complete information on the use and execution of this protocol, please refer to Medendorp et al. (2021).
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Adenosine 5′-triphosphate magnesium salt, ≥95%, bacterial
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Phosphocreatine disodium salt hydrate, ≥97%
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
QX-314, A quaternary lidocaine derivative, permanently charged, and lipophobic.
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis