Merck
  • Lipolysis drives expression of the constitutively active receptor GPR3 to induce adipose thermogenesis.
Lipolysis drives expression of the constitutively active receptor GPR3 to induce adipose thermogenesis.
Cell (2021-05-29)
Olivia Sveidahl Johansen, Tao Ma, Jakob Bondo Hansen, Lasse Kruse Markussen, Renate Schreiber, Laia Reverte-Salisa, Hua Dong, Dan Ploug Christensen, Wenfei Sun, Thorsten Gnad, Iuliia Karavaeva, Thomas Svava Nielsen, Sander Kooijman, Cheryl Cero, Oksana Dmytriyeva, Yachen Shen, Maria Razzoli, Shannon L O'Brien, Eline N Kuipers, Carsten Haagen Nielsen, William Orchard, Nienke Willemsen, Naja Zenius Jespersen, Morten Lundh, Elahu Gosney Sustarsic, Cecilie Mørch Hallgren, Mikkel Frost, Seth McGonigle, Marie Sophie Isidor, Christa Broholm, Oluf Pedersen, Jacob Bo Hansen, Niels Grarup, Torben Hansen, Andreas Kjær, James G Granneman, M Madan Babu, Davide Calebiro, Søren Nielsen, Mikael Rydén, Raymond Soccio, Patrick C N Rensen, Jonas Thue Treebak, Thue Walter Schwartz, Brice Emanuelli, Alessandro Bartolomucci, Alexander Pfeifer, Rudolf Zechner, Camilla Scheele, Susanne Mandrup, Zachary Gerhart-Hines
ABSTRACT
Thermogenic adipocytes possess a therapeutically appealing, energy-expending capacity, which is canonically cold-induced by ligand-dependent activation of β-adrenergic G protein-coupled receptors (GPCRs). Here, we uncover an alternate paradigm of GPCR-mediated adipose thermogenesis through the constitutively active receptor, GPR3. We show that the N terminus of GPR3 confers intrinsic signaling activity, resulting in continuous Gs-coupling and cAMP production without an exogenous ligand. Thus, transcriptional induction of Gpr3 represents the regulatory parallel to ligand-binding of conventional GPCRs. Consequently, increasing Gpr3 expression in thermogenic adipocytes is alone sufficient to drive energy expenditure and counteract metabolic disease in mice. Gpr3 transcription is cold-stimulated by a lipolytic signal, and dietary fat potentiates GPR3-dependent thermogenesis to amplify the response to caloric excess. Moreover, we find GPR3 to be an essential, adrenergic-independent regulator of human brown adipocytes. Taken together, our findings reveal a noncanonical mechanism of GPCR control and thermogenic activation through the lipolysis-induced expression of constitutively active GPR3.
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Sudan Black B, certified by the Biological Stain Commission
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
holo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
D-Pantothenic acid hemicalcium salt, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
(R)-(−)-Isoproterenol, crystalline
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
CL 316,243 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, ≥99%, solid
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Polybrene Infection / Transfection Reagent, A highly efficient method of gene transfer into mammalian cells leveraging infection with retroviral vectors.
Roche
Bovine Serum Albumin Fraction V, heat shock, fatty acid free, from bovine serum
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
(−)-Isoproterenol hydrochloride
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Triacsin C from Streptomyces sp.
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%