Merck
  • AXIN2+ Pericentral Hepatocytes Have Limited Contributions to Liver Homeostasis and Regeneration.
AXIN2+ Pericentral Hepatocytes Have Limited Contributions to Liver Homeostasis and Regeneration.
Cell stem cell (2019-12-24)
Tianliang Sun, Monika Pikiolek, Vanessa Orsini, Sebastian Bergling, Sjoerd Holwerda, Lapo Morelli, Philipp S Hoppe, Lara Planas-Paz, Yi Yang, Heinz Ruffner, Tewis Bouwmeester, Felix Lohmann, Luigi M Terracciano, Guglielmo Roma, Feng Cong, Jan S Tchorz
ABSTRACT
The existence of specialized liver stem cell populations, including AXIN2+ pericentral hepatocytes, that safeguard homeostasis and repair has been controversial. Here, using AXIN2 lineage tracing in BAC-transgenic mice, we confirm the regenerative potential of intestinal stem cells (ISCs) but find limited roles for pericentral hepatocytes in liver parenchyma homeostasis. Liver regrowth following partial hepatectomy is enabled by proliferation of hepatocytes throughout the liver, rather than by a pericentral population. Periportal hepatocyte injury triggers local repair as well as auxiliary proliferation in all liver zones. DTA-mediated ablation of AXIN2+ pericentral hepatocytes transiently disrupts this zone, which is reestablished by conversion of pericentral vein-juxtaposed glutamine synthetase (GS)- hepatocytes into GS+ hepatocytes and by compensatory proliferation of hepatocytes across liver zones. These findings show hepatocytes throughout the liver can upregulate AXIN2 and LGR5 after injury and contribute to liver regeneration on demand, without zonal dominance by a putative pericentral stem cell population.
Sigma-Aldrich
Anti-Albumin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-CYP2E1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial