Merck
  • Pre-microRNA binding aminoglycosides and antitumor drugs as inhibitors of Dicer catalyzed microRNA processing.
Pre-microRNA binding aminoglycosides and antitumor drugs as inhibitors of Dicer catalyzed microRNA processing.
Bioorganic & medicinal chemistry letters (2012-01-20)
Mohitosh Maiti, Koen Nauwelaerts, Piet Herdewijn
ABSTRACT
Over-expressions of miRNAs are being increasingly linked with many diseases including different types of cancer. In this study, the role of some known small molecular therapeutics has been investigated for their ability to bind with the pre-miRNA target (hsa-mir-155) and thereby to interfere with the Dicer catalyzed miRNA processing. Potential binding and inhibition effects have been demonstrated by some of these analogs. They can be used as leads for further development of potent small molecular miRNA-antagonists.
Product Description
Supelco
4-Epianhydrotetracycline hydrochloride, VETRANAL®, analytical standard
Supelco
4-Epitetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Demeclocycline hydrochloride, powder or crystals
Supelco
Anhydrotetracycline hydrochloride, VETRANAL®, analytical standard