Merck
  • Eubacterium rectale contributes to colorectal cancer initiation via promoting colitis.
Eubacterium rectale contributes to colorectal cancer initiation via promoting colitis.
Gut pathogens (2021-01-14)
Yijia Wang, Xuehua Wan, Xiaojing Wu, Chunze Zhang, Jun Liu, Shaobin Hou
ABSTRACT
Inflammatory bowel disease caused by microbial dysbiosis is an important factor contributing to colorectal cancer (CRC) initiation. The 'driver-passenger' model in human gut microbial dysbiosis suggests that 'driver' bacteria may colonize with low relative abundance on tumor site but persistently induce chronic change in normal intestinal epithelium and initiate CRC. They are gradually replaced by 'passenger' bacteria later on, due to their low adaptability to the on-tumor site niche. To reveal site-specific bacterial taxon markers in CRC patients, we analyzed the gut mucosal microbiome of 75 paired samples of on-tumor and tumor-adjacent sites, 75 off-tumor sites, and 26 healthy controls. Linear discriminant analysis of relative abundance profiles revealed unique bacterial taxon distribution correlated with specific tumor sites, with Eubacterium having the distribution characteristic of potential driver bacteria. We further show that Eubacterium rectale endotoxin activates the transcription factor NF-κΒ, which regulates multiple aspects of innate and adaptive immune responses in normal colon epithelial cells. Unlike the 'passenger' bacterium Fusobacterium nucleatum, E. rectale promotes dextran sodium sulfate-induced colitis in Balb/c mice. Our findings reveal that E. rectale functions as a 'driver' bacterium and contributes to cancer initiation via promoting inflammation.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
ANTI-IKB ALPHA (IKBA) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Histone H3 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-NF-κB p65 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, FITC conjugate, 2 mg/mL, Chemicon®