Merck
Interleukin-24 protects against liver injury in mouse models.
EBioMedicine (2021-01-29)
Hsiao-Hsuan Wang, Jian-Hao Huang, Min-Hao Sue, Wei-Chih Ho, Yu-Hsiang Hsu, Kung-Chao Chang, Ming-Shi Chang
ABSTRACT
Interleukin-24 (IL-24) binds to two kinds of receptor complexes, namely IL-20R1/IL-20R2 and IL-20R2/IL-22R1, which are also bound by IL-20. IL-20 plays a detrimental role in liver fibrosis. Due to the sharing of receptor complexes, we aimed to determine whether IL-24 also participates in liver fibrosis. Clinical biopsy specimens from various stages of liver fibrosis were used to analyze IL-24 expression. IL-24 protein was administered to mice with thioacetamide (TAA)-induced liver injury. The direct effects of IL-24 on mouse primary hepatocytes or hepatic stellate cells (HSCs) were analyzed. Wild-type, IL-20R1-, and IL20R2-deficient mice were used to establish a model of acute TAA-induced liver injury. Among patients with more severe liver fibrosis, there was a reduced IL-24/IL-20 ratio. Administration of IL-24 protein protected mice from TAA-induced liver injury and reduction of liver inflammation by antioxidant effects. IL-24 protected hepatocytes from TAA-induced apoptosis and prevented liver fibrosis through the inhibition of the HSCs activation. The protective role of IL-24 acted on liver cells were mainly IL-20R1-independent. IL-20R2-deficient mice exhibited more severe liver injury upon TAA treatment, thus confirming the protective role of IL-24. IL-24 plays a key protective role in the progression of liver injury and has therapeutic potential for treating liver injuries. This work was supported by the Ministry of Science and Technology of Taiwan (MOST 106-2320-B-006-024) and Taiwan Liver Disease Prevention & Treatment Research Foundation.
Sigma-Aldrich
Thioacetamide, ACS reagent, ≥99.0%
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
HSC-T6 Rat Hepatic Stellate Cell Line, The HSC-T6 rat hepatic stellate cell line is a valuable cell model for studies of retinoid metabolism based on their similar retinoid phenotype as primary cells.