Merck
  • Antiallergic potential on RBL-2H3 cells of some phenolic constituents of Zingiber officinale (ginger).
Antiallergic potential on RBL-2H3 cells of some phenolic constituents of Zingiber officinale (ginger).
Journal of natural products (2009-03-11)
Bing-Hung Chen, Pei-Yu Wu, Kuan-Ming Chen, Tzu-Fun Fu, Hui-Min Wang, Chung-Yi Chen
ABSTRACT
In the present study on five pure phenolic compounds (1-5) isolated from the rhizomes of Zingiber officinale (ginger) and investigated for their antiallergic potency, rat basophilic leukemia (RBL-2H3) cells were incubated with these compounds and the release of beta-hexosaminidase was measured kinetically. The data obtained suggest that ginger rhizomes harbor potent compounds capable of inhibiting allergic reactions and may be useful for the treatment and prevention of allergic diseases.
Product Description
Supelco
[6]-Gingerol, analytical standard
Supelco
[10]-Gingerol, analytical standard
Sigma-Aldrich
[6]-Gingerol, ≥98% (HPLC)