Merck
  • Assessment of pregnancy-associated glycoprotein profile in milk for early pregnancy diagnosis in goats.
Assessment of pregnancy-associated glycoprotein profile in milk for early pregnancy diagnosis in goats.
Animal bioscience (2020-02-15)
Shiva Pratap Singh, Ramachandran Natesan, Nandini Sharma, Anil Kumar Goel, Manoj Kumar Singh, Suresh Dinkar Kharche
ABSTRACT
This study was conducted to assess the level of pregnancy-associated glycoprotein (PAG) in whole and skim milk samples, and its suitability for early pregnancy diagnosis in goats. A two-step sandwich enzyme-linked immunosorbent assay (ELISA) system for estimation of milk PAG was developed and validated, which employed caprine-PAG specific polyclonal antisera. Whole and skim milk samples (n = 210 each) from fifteen multiparous goats were collected on alternate days from d 10 to d 30, and thereafter weekly till d 51 postmating. PAG levels in milk samples were estimated by ELISA and the pregnancies were confirmed at d40 post-mating by transrectal ultrasonography (TRUS). The level of PAG in whole and skim milk samples of both pregnant and nonpregnant goats remained below the threshold values until d 24 after mating. Thereafter, PAG concentration in whole and skim milk increased steadily in pregnant goats, whereas it continued below the threshold in non-pregnant does. The PAG profiles in whole and skim milk of pregnant goats were almost similar and exhibited strong positive relationship (r = 0.891; p<0.001). Day 26 post-mating was identified as the first time-point for significantly (p<0.05) higher milk PAG concentration in pregnant goats than to non-pregnant goats. When compared to TRUS examination for pregnancy diagnosis, the accuracy and specificity of PAG ELISA using whole and skim milk samples were 94.5% and 95.4%; and 95.3% and 100%, respectively. The high values of area-under-curve (0.904 [whole milk] and 0.922 [skim milk]), demonstrate outstanding discrimination ability of the milk assays. Among the sampling dates chosen, d 37 post-mating was identified as the best suitable time point for collection of milk samples to detect pregnancy in goats. The PAG concentration in whole and skim milk of goats collected between days 26 and 51 post-breeding can be used for the accurate prediction of pregnancy and may be useful for assisting management decisions in goat flocks.
Sigma-Aldrich
Mix-n-Stain Biotin Antibody Labeling Kit (50-100μg)
Sigma-Aldrich
Avidin–Peroxidase, lyophilized powder