Merck
  • Pleckstrin-2 is essential for erythropoiesis in β-thalassemic mice, reducing apoptosis and enhancing enucleation.
Pleckstrin-2 is essential for erythropoiesis in β-thalassemic mice, reducing apoptosis and enhancing enucleation.
Communications biology (2021-05-05)
Maria Feola, Andrea Zamperone, Daniel Moskop, Huiyong Chen, Carla Casu, Dechen Lama, Julie Di Martino, Mansour Djedaini, Luena Papa, Marc Ruiz Martinez, Tenzin Choesang, Jose Javier Bravo-Cordero, Matthew MacKay, Paul Zumbo, Nathan Brinkman, Charles S Abrams, Stefano Rivella, Shilpa Hattangadi, Christopher E Mason, Ronald Hoffman, Peng Ji, Antonia Follenzi, Yelena Z Ginzburg
ABSTRACT
Erythropoiesis involves complex interrelated molecular signals influencing cell survival, differentiation, and enucleation. Diseases associated with ineffective erythropoiesis, such as β-thalassemias, exhibit erythroid expansion and defective enucleation. Clear mechanistic determinants of what make erythropoiesis effective are lacking. We previously demonstrated that exogenous transferrin ameliorates ineffective erythropoiesis in β-thalassemic mice. In the current work, we utilize transferrin treatment to elucidate a molecular signature of ineffective erythropoiesis in β-thalassemia. We hypothesize that compensatory mechanisms are required in β-thalassemic erythropoiesis to prevent apoptosis and enhance enucleation. We identify pleckstrin-2-a STAT5-dependent lipid binding protein downstream of erythropoietin-as an important regulatory node. We demonstrate that partial loss of pleckstrin-2 leads to worsening ineffective erythropoiesis and pleckstrin-2 knockout leads to embryonic lethality in β-thalassemic mice. In addition, the membrane-associated active form of pleckstrin-2 occurs at an earlier stage during β-thalassemic erythropoiesis. Furthermore, membrane-associated activated pleckstrin-2 decreases cofilin mitochondrial localization in β-thalassemic erythroblasts and pleckstrin-2 knockdown in vitro induces cofilin-mediated apoptosis in β-thalassemic erythroblasts. Lastly, pleckstrin-2 enhances enucleation by interacting with and activating RacGTPases in β-thalassemic erythroblasts. This data elucidates the important compensatory role of pleckstrin-2 in β-thalassemia and provides support for the development of targeted therapeutics in diseases of ineffective erythropoiesis.
Sigma-Aldrich
Anti-VDAC1 Antibody, clone N152B/23, clone N152B/23, from mouse
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
NSC23766 trihydrochloride, ≥97% (HPLC)