Merck
  • Acquiring Metastatic Competence by Oral Squamous Cell Carcinoma Cells Is Associated with Differential Expression of α-Tubulin Isoforms.
Acquiring Metastatic Competence by Oral Squamous Cell Carcinoma Cells Is Associated with Differential Expression of α-Tubulin Isoforms.
Journal of oncology (2012-06-22)
Becky Lou, David Engler, William Dubinsky, Jean Wu, Nadarajah Vigneswaran
ABSTRACT
We performed comparative global proteomics analyses of patient-matched primary (686Tu) and metastatic (686Ln) OSCC cells. The metastatic OSCC 686Ln cells showed greater in vitro migratory/invasive potential and distinct cell shape from their parental primary 686Tu cells. Ettan DIGE analysis revealed 1316 proteins spots in both cell lines with >85% to be quantitatively similar (<2 folds) between the two cell lines. However, two protein spots among four serial spots were highly dominant in 686Ln cells. Mass spectrometry sequencing demonstrated all four spots to be α-tubulin isotypes. Further analysis showed no significant quantitative difference in the α-tubulin between the two cell lines either at mRNA or protein levels. Thus, two distinct isoforms of α-tubulin, probably due to posttranslational modification, were associated with metastatic 686Ln cells. Immunofluorescence demonstrated remarkable differences in the cytosolic α-tubulin distribution patterns between the two cells. In 686Tu cells, α-tubulin proteins formed a normal network composed of filaments. In contrast, α-tubulin in 686Ln cells exhibited only partial cytoskeletal distribution with the majority of the protein diffusely distributed within the cytosol. Since α-tubulin is critical for cell shape and mobility, our finding suggests a role of α-tubulin isoforms in acquisition of metastatic phenotype and represents potential target for therapeutic intervention.
Sigma-Aldrich
Anti-BAG3 Antibody, clone AC-1, clone AC-1, from mouse
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid