Merck
  • Polymeric nanoparticles for selective protein recognition by using thiol-ene miniemulsion photopolymerization.
Polymeric nanoparticles for selective protein recognition by using thiol-ene miniemulsion photopolymerization.
Journal of biomaterials science. Polymer edition (2020-07-10)
Merve Yaşar, Fulya Dal Yöntem, M Vezir Kahraman, Nilhan Kayaman Apohan, Zerrin Aktaş, Mustafa Oral Öncül, Handan Akçakaya
ABSTRACT
The fabrication of molecularly imprinted nanoparticles (MIP-NPs) specific for myoglobin by using thiol-ene photopolymerization in miniemulsion was described. Allyl derivatives of phenylalanine as a functional monomer was synthesized and copolymerized with acrylic monomers via miniemulsion polymerization to produce NIP-NPs with approximately 74 nm number average particle diameter. FTIR and 1H-NMR analysis confirmed the synthesis of functional monomer. MIP-NPs were prepared in the existence of myoglobin as a template protein. Morphological investigations exhibited that the particle size of the MIP-NPs, increased compared to the corresponding NIPs and the mean particle diameter by number was measured as 141 nm with narrow distribution. NIP-NPs that were polymerized without myoglobin were found to have less affinity to the target protein. In addition, the rebinding ability of MIP-NPs was much bigger than that of the corresponding NIPs. ELISA results showed that MIPs interact particularly with the myoglobin and show little affinity for BSA in competitive binding experiments.HighlightsAllyl N,N-diallyl phenylalaninate was synthesized as a functional monomer.Imprinted nanoparticles were prepared by using thiol-ene photopolymerization in miniemulsion.The nanoparticles were 141 nm with narrow size distribution.The imprinted nanoparticles showed selectivity toward myoglobin.
Sigma-Aldrich
N-[3-(Dimethylamino)propyl]methacrylamide, 99%, contains MEHQ as inhibitor
Sigma-Aldrich
1-Hydroxycyclohexyl phenyl ketone, 99%
Sigma-Aldrich
Pentaerythritol tetrakis(3-mercaptopropionate), >95%
Sigma-Aldrich
N-Hydroxyethyl acrylamide, contains 1,000 ppm monomethyl ether hydroquinone as stabilizer, 97%