Merck
  • TRIM37 prevents formation of centriolar protein assemblies by regulating Centrobin.
TRIM37 prevents formation of centriolar protein assemblies by regulating Centrobin.
eLife (2021-01-26)
Fernando R Balestra, Andrés Domínguez-Calvo, Benita Wolf, Coralie Busso, Alizée Buff, Tessa Averink, Marita Lipsanen-Nyman, Pablo Huertas, Rosa M Ríos, Pierre Gönczy
ABSTRACT
TRIM37 is an E3 ubiquitin ligase mutated in Mulibrey nanism, a disease with impaired organ growth and increased tumor formation. TRIM37 depletion from tissue culture cells results in supernumerary foci bearing the centriolar protein Centrin. Here, we characterize these centriolar protein assemblies (Cenpas) to uncover the mechanism of action of TRIM37. We find that an atypical de novo assembly pathway can generate Cenpas that act as microtubule-organizing centers (MTOCs), including in Mulibrey patient cells. Correlative light electron microscopy reveals that Cenpas are centriole-related or electron-dense structures with stripes. TRIM37 regulates the stability and solubility of Centrobin, which accumulates in elongated entities resembling the striped electron dense structures upon TRIM37 depletion. Furthermore, Cenpas formation upon TRIM37 depletion requires PLK4, as well as two parallel pathways relying respectively on Centrobin and PLK1. Overall, our work uncovers how TRIM37 prevents Cenpas formation, which would otherwise threaten genome integrity.
Sigma-Aldrich
MISSION® esiRNA, targeting human CNTROB
Sigma-Aldrich
MISSION® esiRNA, targeting human TRIM37
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, purified from hybridoma cell culture
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Sigma-Aldrich
Anti-CDK5RAP2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-CEP152 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SPICE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CEP170 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human PLK4
Sigma-Aldrich
MISSION® esiRNA, targeting human CENPJ
Sigma-Aldrich
MISSION® esiRNA, targeting human SASS6
Sigma-Aldrich
MISSION® esiRNA, targeting human STIL