Merck
  • Loss of CD36 impairs hepatic insulin signaling by enhancing the interaction of PTP1B with IR.
Loss of CD36 impairs hepatic insulin signaling by enhancing the interaction of PTP1B with IR.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-02-27)
Ping Yang, Han Zeng, Wei Tan, Xiaoqing Luo, Enze Zheng, Lei Zhao, Li Wei, Xiong Z Ruan, Yao Chen, Yaxi Chen
ABSTRACT
A contradictory role of CD36 in insulin resistance was found to be related to the nutrient state. Here, we examined that the physiological functions of CD36 in insulin signal transduction in mice fed a low-fat diet. CD36 deficiency led to hepatic insulin resistance and decreased insulin-stimulated tyrosine phosphorylation of insulin receptor β (IRβ) in mice fed a low-fat diet. The ability of insulin to bind with IR did not differ between WT and CD36-deficient hepatocytes. CD36 formed a complex with IRβ and dissociation of CD36/Fyn complex or inhibition of Fyn only partially reversed the effects of CD36 on hepatic insulin signaling. Furthermore, we found that CD36 deficiency led to abnormally increased hepatic protein-tyrosine phosphatase 1B (PTP1B) expression and enhanced PTP1B and IR interactions, which contributed to the decreased insulin signaling and disordered glucose metabolism. In addition, increased endoplasmic reticulum (ER) stress was found in the livers of the CD36-deficient mice, while inhibited ER stress normalized the PTP1B expression and restored insulin signaling in the CD36-deficient mice. Our findings suggest that the loss of CD36 impairs hepatic insulin signaling by enhancing the PTP1B/IR interaction that is induced by ER stress, indicating a possible critical step in the progression of hepatic insulin resistance.
Sigma-Aldrich
Anti-FOXO1 Antibody, clone 2H8.2, clone 2H8.2, from mouse
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-IRS1 Antibody, clone AW58, rabbit monoclonal, culture supernatant, clone AW58, Upstate®
Sigma-Aldrich
Claramine trifluoroacetate salt, ≥98% (HPLC)