Merck
  • Structures of PGAM5 Provide Insight into Active Site Plasticity and Multimeric Assembly.
Structures of PGAM5 Provide Insight into Active Site Plasticity and Multimeric Assembly.
Structure (London, England : 1993) (2017-06-27)
Apirat Chaikuad, Panagis Filippakopoulos, Sean R Marcsisin, Sarah Picaud, Martin Schröder, Shiori Sekine, Hidenori Ichijo, John R Engen, Kohsuke Takeda, Stefan Knapp
ABSTRACT
PGAM5 is a mitochondrial membrane protein that functions as an atypical Ser/Thr phosphatase and is a regulator of oxidative stress response, necroptosis, and autophagy. Here we present several crystal structures of PGAM5 including the activating N-terminal regulatory sequences, providing a model for structural plasticity, dimerization of the catalytic domain, and the assembly into an enzymatically active dodecameric form. Oligomeric states observed in structures were supported by hydrogen exchange mass spectrometry, size-exclusion chromatography, and analytical ultracentrifugation experiments in solution. We report that the catalytically important N-terminal WDPNWD motif acts as a structural integrator assembling PGAM5 into a dodecamer, allosterically activating the phosphatase by promoting an ordering of the catalytic loop. Additionally the observed active site plasticity enabled visualization of essential conformational rearrangements of catalytic elements. The comprehensive biophysical characterization offers detailed structural models of this key mitochondrial phosphatase that has been associated with the development of diverse diseases.
Sigma-Aldrich
Rosetta(DE3) Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
Roche
Anti-HA High Affinity, from rat IgG1
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution