Merck
Importance of phosphatidylcholine on the chloroplast surface.
Progress in lipid research (2016-11-23)
César Botella, Juliette Jouhet, Maryse A Block
ABSTRACT
In plant cells, phosphatidylcholine (PC) is a major glycerolipid of most membranes but practically lacking from the plastid internal membranes. In chloroplasts, PC is absent from the thylakoids and the inner envelope membrane. It is however the main component of the outer envelope membrane, where it exclusively distributes in the outer monolayer. This unique distribution is likely related with operational compartmentalization of plant lipid metabolism. In this review, we summarize the different mechanisms involved in homeostasis of PC in plant cells. The specific origin of chloroplast PC is examined and the involvement of the P4-ATPase family of phospholipid flippases (ALA) is considered with a special attention to the recently reported effect of the endoplasmic reticulum-localized ALA10 on modification of chloroplast PC desaturation. The different possible roles of chloroplast PC are then discussed and analyzed in consideration of plant physiology.
Product Description
Avanti
14:0-12:0 NBD PC, Avanti Research - A Croda Brand 810123P, powder
Avanti
18:1-06:0 NBD PC, Avanti Research - A Croda Brand
Avanti
16:0-Pyr PC, Avanti Research - A Croda Brand 810197P, powder
Avanti
TopFluor TMR PC, Avanti Research - A Croda Brand
Avanti
18:1-12:0 NBD PC, Avanti Research - A Croda Brand 810133C
Avanti
16:0-12:0 NBD PC, Avanti Research - A Croda Brand 810131C
Avanti
14:0-06:0 NBD PC, Avanti Research - A Croda Brand 810122P, powder
Avanti
16:0-12:0 NBD PC, Avanti Research - A Croda Brand 810131P, powder
Avanti
TopFluor TMR PC, Avanti Research - A Croda Brand 810180C