Merck
Novel enzymatic method for assaying Lp-PLA2 in serum.
Clinica chimica acta; international journal of clinical chemistry (2018-03-20)
Saki Yamaura, Shin-Ichi Sakasegawa, Emisa Koguma, Shigeru Ueda, Yuzo Kayamori, Daisuke Sugimori, Ken Karasawa
ABSTRACT
Measurement of lipoprotein-associated phospholipase A2 (Lp-PLA2) can be used as an adjunct to traditional cardiovascular risk factors for identifying individuals at higher risk of cardiovascular events. This can be performed by quantification of the protein concentration using an ELISA platform or by measuring Lp-PLA2 activity using platelet-activating factor (PAF) analog as substrate. Here, an enzymatic Lp-PLA2 activity assay method using 1-O-Hexadecyl-2-acetyl-rac-glycero-3-phosphocholine (rac C16 PAF) was developed. The newly revealed substrate specificity of lysoplasmalogen-specific phospholipase D (lysophospholipase D (LysoPLD)) was exploited. Lp-PLA2 hydrolyzes 1-O-Hexadecyl-2-acetyl-sn-glycero-3-phosphocholine (C16 PAF) to 1-O-Hexadecyl-2-hydroxy-sn-glycero-3-phosphocholine (LysoPAF). LysoPLD acted on LysoPAF, and the hydrolytically released choline was detected by choline oxidase. Regression analysis of Lp-PLA2 activity measured by the enzymatic Lp-PLA2 activity assay vs. two chemical Lp-PLA2 activity assays, i.e. LpPLA2 FS and PLAC® test, and ELISA, gave the following correlation coefficients: 0.990, 0.893 and 0.785, respectively (n = 30). Advantages of this enzymatic Lp-PLA2 activity assay compared with chemical Lp-PLA2 methods include the following; (i) only requires two reagents enabling a simple two-point linear calibration method with one calibrator (ii) no need for inhibitors of esterase-like activity in serum.
Product Description
Avanti
C18-02:0 PC, Avanti Research - A Croda Brand 878114P, powder
Avanti
C16-02:0 PC, Avanti Research - A Croda Brand 878110P, powder
Avanti
C16-02:0 PC, Avanti Research - A Croda Brand 878110C
Avanti
18:1 PEG2000 PE, Avanti Research - A Croda Brand 880130P, powder
Avanti
C16-18:1 PC, Avanti Research - A Croda Brand 878112P, powder