Merck
  • Upregulation of ATG7 attenuates motor neuron dysfunction associated with depletion of TARDBP/TDP-43.
Upregulation of ATG7 attenuates motor neuron dysfunction associated with depletion of TARDBP/TDP-43.
Autophagy (2019-06-27)
Aneesh Donde, Mingkuan Sun, Yun Ha Jeong, Xinrui Wen, Jonathan Ling, Sophie Lin, Kerstin Braunstein, Shuke Nie, Sheng Wang, Liam Chen, Philip C Wong
ABSTRACT
A shared neuropathological hallmark in amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD) is nuclear clearance and cytoplasmic aggregation of TARDBP/TDP-43 (TAR DNA binding protein). We previously showed that the ability of TARDBP to repress nonconserved cryptic exons was impaired in brains of patients with ALS and FTD, suggesting that its nuclear depletion contributes to neurodegeneration. However, the critical pathways impacted by the failure to repress cryptic exons that may contribute to neurodegeneration remain undefined. Here, we report that transcriptome analysis of TARDBP-deficient neurons revealed downregulation of ATG7, a critical gene required for macroautophagy/autophagy. Mouse and Drosophila models lacking TARDBP/TBPH in motor neurons exhibiting age-dependent neurodegeneration and motor deficits showed reduction of ATG7 and accumulation of SQSTM1/p62 inclusions. Importantly, genetic upregulation of the autophagy pathway improved motor function and survival in TBPH-deficient flies. Together with our observation that ATG7 is reduced in ALS-FTD brain tissues, these findings identify the autophagy pathway as one key effector of nuclear depletion of TARDBP that contributes to neurodegeneration. We thus suggest that the autophagy pathway is a therapeutic target for ALS-FTD and other disorders exhibiting TARDBP pathology.Abbreviations: ALS: amyotrophic lateral sclerosis; ANOVA: analysis of variance; ChAT: choline acetyltransferase; CTSD: cathepsin D; FTD: frontotemporal dementia; LAMP1: lysosomal associated membrane protein 1; NMJ: neuromuscular junction; RBFOX3/NeuN: RNA binding fox-1 homolog 3; SQSTM1: sequestosome 1; TARDBP/TDP-43: TAR DNA binding protein 43.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
Toluidine Blue, 8.74% (ZN (THEORY)), for microscopy (Hist., Vit.)
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Supelco
Isopentane Solution, Pharmaceutical Secondary Standard; Certified Reference Material