Merck
  • Developmental lead (Pb)-induced deficits in hippocampal protein translation at the synapses are ameliorated by ascorbate supplementation.
Developmental lead (Pb)-induced deficits in hippocampal protein translation at the synapses are ameliorated by ascorbate supplementation.
Neuropsychiatric disease and treatment (2018-12-21)
Faraz Ahmad, Mohammad Salahuddin, Khaldoon Alsamman, Abdulaziz A AlMulla, Khaled F Salama
ABSTRACT
Lead (Pb) is a persistent environmental neurotoxin and its exposure even in minute quantities has been known to induce neuronal defects. The immature brain is singularly sensitive to Pb neurotoxicity, and its exposure during development has permanent detrimental effects on the brain developmental trajectory and neuronal signaling and plasticity, culminating into compromises in the cognitive and behavioral attributes which persists even later in adulthood. Several molecular pathways have been implicated in the Pb-mediated disruption of neuronal signaling, including elevated oxidative stress, alterations in neurotransmitter biology, and mitochondrial dysfunction. Nevertheless, the neuronal targets and biochemical pathways underlying these Pb-mediated alterations in synaptic development and function have not been completely deduced. In this respect, recent studies have shown that synaptic signaling and its maintenance and plasticity are critically dependent on localized de novo protein translation at the synaptic terminals. The present study hence aimed to assess the alterations in the synapse-specific translation induced by developmental Pb exposure. To this end, in vitro protein translation rate was analyzed in the hippocampal synaptoneurosomal fractions of rat pups pre- and postnatally exposed to Pb using a puromycin incorporation assay. Moreover, we evaluated the therapeutic effects of ascorbic acid supplementation against Pb-induced deficits in synapse-localized protein translation. We observed a significant loss in the rates of de novo protein translation in synaptoneurosomes of Pb-exposed pups compared to age-matched control pups. Interestingly, ascorbate supplementation lead to an appreciable recovery in Pb-induced translational deficits. Moreover, the deficit in activity-dependent synaptic protein translation was found to correlate significantly with the increase in the blood Pb levels. Dysregulation of synapse-localized de novo protein translation is a potentially critical determinant of Pb-induced synaptic dysfunction and the consequent deficits in behavioral, social, and psychological attributes of the organisms. In addition, our study establishes ascorbate supplementation as a key ameliorative agent against Pb-induced neurotoxicity.
Sigma-Aldrich
Anti-Puromycin Antibody, clone 17H1, clone 17H1, from rat
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)