Merck
  • Brain pharmacology of intrathecal antisense oligonucleotides revealed through multimodal imaging.
Brain pharmacology of intrathecal antisense oligonucleotides revealed through multimodal imaging.
JCI insight (2019-10-18)
Curt Mazur, Berit Powers, Kenneth Zasadny, Jenna M Sullivan, Hemi Dimant, Fredrik Kamme, Jacob Hesterman, John Matson, Michael Oestergaard, Marc Seaman, Robert W Holt, Mohammed Qutaish, Ildiko Polyak, Richard Coelho, Vijay Gottumukkala, Carolynn M Gaut, Marc Berridge, Nazira J Albargothy, Louise Kelly, Roxana O Carare, Jack Hoppin, Holly Kordasiewicz, Eric E Swayze, Ajay Verma
ABSTRACT
Intrathecal (IT) delivery and pharmacology of antisense oligonucleotides (ASOs) for the CNS have been successfully developed to treat spinal muscular atrophy. However, ASO pharmacokinetic (PK) and pharmacodynamic (PD) properties remain poorly understood in the IT compartment. We applied multimodal imaging techniques to elucidate the IT PK and PD of unlabeled, radioactively labeled, or fluorescently labeled ASOs targeting ubiquitously expressed or neuron-specific RNAs. Following lumbar IT bolus injection in rats, all ASOs spread rostrally along the neuraxis, adhered to meninges, and were partially cleared to peripheral lymph nodes and kidneys. Rapid association with the pia and arterial walls preceded passage of ASOs across the glia limitans, along arterial intramural basement membranes, and along white-matter axonal bundles. Several neuronal and glial cell types accumulated ASOs over time, with evidence of probable glial accumulation preceding neuronal uptake. IT doses of anti-GluR1 and anti-Gabra1 ASOs markedly reduced the mRNA and protein levels of their respective neurotransmitter receptor protein targets by 2 weeks and anti-Gabra1 ASOs also reduced binding of the GABAA receptor PET ligand 18F-flumazenil in the brain over 4 weeks. Our multimodal imaging approaches elucidate multiple transport routes underlying the CNS distribution, clearance, and efficacy of IT-dosed ASOs.
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, from chicken, purified by affinity chromatography