Merck
Chicken ghrelin: purification, cDNA cloning, and biological activity.
Endocrinology (2002-08-24)
Hiroyuki Kaiya, Serge Van Der Geyten, Masayasu Kojima, Hiroshi Hosoda, Yasuo Kitajima, Masaru Matsumoto, Sofie Geelissen, Veerle M Darras, Kenji Kangawa
ABSTRACT
In this study, we report the purification, cDNA cloning, and characterization of the novel growth hormone-releasing peptide, ghrelin, in the chicken (Gallus gallus). Chicken ghrelin is composed of 26 amino acids (GSSFLSPTYKNIQQQKDTRKPTARLH) and possesses 54% sequence identity with human ghrelin. The serine residue at position 3 (Ser(3)) is conserved between the chicken and mammalian species, as its acylation by either n-octanoic or n-decanoic acid. Chicken ghrelin mRNA is predominantly expressed in the stomach, where it is present in the proventriculus but absent in the gizzard. Using RT-PCR analysis, low levels of expression were also detectable in brain, lung, and intestine. Administration of chicken ghrelin increases plasma GH levels in both rats and chicks, with a potency similar to that of rat or human ghrelin. In addition, chicken ghrelin also increases plasma corticosterone levels in growing chicks at a lower dose than in mammals. The present results indicate that the stimulatory effect of ghrelin on GH secretion is evolutionarily conserved, whereas its effect on adrenal function seems to be unique in the chicken.
Sigma-Aldrich
ReadyMix Taq PCR Reaction Mix, with MgCl2
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Supelco
Decanoic acid, analytical standard
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥98.0%