Merck
  • Simultaneous Time-concentration Analysis of Soman and VX Adducts to Butyrylcholinesterase and Albumin by LC-MS-MS.
Simultaneous Time-concentration Analysis of Soman and VX Adducts to Butyrylcholinesterase and Albumin by LC-MS-MS.
Journal of analytical toxicology (2018-04-05)
Jin Young Lee, Changhwan Kim, Yong Han Lee
ABSTRACT
A sensitive method for the purification and determination of two protein adducts, organophosphorus (OP)-BChE and OP-albumin adducts, in a single sample using a simultaneous sample preparation method was developed and validated using liquid chromatography-tandem mass spectrometry. First, we isolated O-ethyl S-2-diisopropylaminoethyl methyl phosphonothiolate (VX) and O-pinacolyl methylphosphonofluoridate (soman, GD)-BChE adducts using an immunomagnetic separation (IMS) method and the HiTrap™ Blue affinity column was subsequently used to isolate and purify VX and GD-albumin adducts from the plasma of rhesus monkeys exposed to nerve agents. Additionally, we examined the time-concentration profiles of two biomarkers, VX and GD-nonapeptides and VX and GD-tyrosines, derived from OP-BChE and OP-albumin adducts up to 8 weeks after exposure. Based on the results, we determined that VX and GD-tyrosine is more suitable than VX and GD-nonapeptide as a biomarker owing to its longevity. This integrated approach is expected to be applicable for the quantification of other OP-BChE and OP-albumin adducts in human plasma, thus serving as a potential generic assay for exposure to nerve agents.
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution
Sigma-Aldrich
Phosphate Buffered Saline with 0.05% TWEEN® 20, pH 7.4
Sigma-Aldrich
Dess-Martin periodinane, 97%
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Triethanolamine Buffer solution