Vitamin K2
Synonym(s):
Menaquinone 4, Menatetrenone
Empirical Formula (Hill Notation):
C31H40O2
CAS Number:
Molecular Weight:
444.65
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing