
Tri-tert-butylphosphine solution

Synonym(s):
P(t-Bu)3
Empirical Formula (Hill Notation):
C12H27P
CAS Number:
Molecular Weight:
202.32
Products
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing