
Trimethylolpropane ethoxylate

Synonym(s):
3 arm PEG, 3 arm PEO, Poly(ethylene oxide), 3-arm, hydroxy terminated, Polyethylene glycol trimethylolpropane ether, Polyethylene glycol trimethylolpropane triether, Trimethylolpropane ethoxylate (20/3 EO/OH), Trimethylolpropane ethoxylate (4.6 EO/OH), Trimethylolpropane ethoxylate (4/15 EO/OH)
Linear Formula:
(C2H4O)n(C2H4O)n(C2H4O)nC6H14O
CAS Number:
Products
Physical Form
Showing 1-2 of 2
Compare
Product Number
Description
Pricing