
Trifluoromethanesulfonic acid-d

Synonym(s):
Deuterotrifluormethansulfonic acid, Triflic acid-D
Linear Formula:
CF3SO3D
CAS Number:
Molecular Weight:
151.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing