
Triethyl phosphite

Synonym(s):
P(EtO)3, P(OEt)3, Triethylphosphite
Linear Formula:
(C2H5O)3P
CAS Number:
Molecular Weight:
166.16
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing