
Transforming Growth Factor-α human

Synonym(s):
TGF-α
Empirical Formula (Hill Notation):
C239H348N70O72S6
CAS Number:
Molecular Weight:
5546.14
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing