Skip to Content
Merck
(−)-Tetramisole hydrochloride ≥99% (GC)

(−)-Tetramisole hydrochloride

(−)-Tetramisole hydrochloride ≥99% (GC)
Synonym(s):
(S)-(−)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride, L(−)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, Levamisole hydrochloride
Empirical Formula (Hill Notation):
C11H12N2S · HCl
CAS Number:
Molecular Weight:
240.75
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Mode of Action
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing