
TCPP, mixture of isomers

Empirical Formula (Hill Notation):
C9H18Cl3O4P
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing