![Sulconazole Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/103/399/0d7d1d34-d304-4e53-88ac-1a5367c4f6eb/640/0d7d1d34-d304-4e53-88ac-1a5367c4f6eb.jpg)
Sulconazole Related Compound A
![Sulconazole Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/103/399/0d7d1d34-d304-4e53-88ac-1a5367c4f6eb/640/0d7d1d34-d304-4e53-88ac-1a5367c4f6eb.jpg)
Synonym(s):
1-[2-(4-Chlorobenzyl)sulfinyl-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole nitrate
Empirical Formula (Hill Notation):
C18H15Cl3N2OS · HNO3
CAS Number:
Molecular Weight:
476.76
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2