
Sodium phosphate tribasic dodecahydrate

Synonym(s):
TSP, Trisodium phosphate (tert) dodecahydrate, Trisodium phosphate dodecahydrate
Linear Formula:
Na3PO4 · 12H2O
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing