Skip to Content
Merck
Sodium Phosphate OmniPur® Grade, >= 98.0%

Sodium Phosphate

Sodium Phosphate OmniPur® Grade, >= 98.0%
Synonym(s):
Sodium phosphate dibasic heptahydrate, Disodium hydrogen phosphate heptahydrate, Disodium phosphate
Linear Formula:
Na2HPO4 · 7H2O
CAS Number:
Molecular Weight:
268.07

Products

Brand
Color
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing