
S-Allyl-L-cysteine

Synonym(s):
L-Deoxyalliin, S-allyl-cysteine, S-allylcysteine, SAC
Empirical Formula (Hill Notation):
C6H11NO2S
CAS Number:
Molecular Weight:
161.22
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing