
Protodioscin

Empirical Formula (Hill Notation):
C51H84O22
CAS Number:
Molecular Weight:
1049.20
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing