
Prostaglandin D2

Synonym(s):
(5Z,9α,13E,15S)-9,15-Dihydroxy-11-oxoprosta-5,13-dien-1-oic-acid, PGD2, Prostaglandin D2
Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing