
Potassium Phosphate Monobasic solution

Synonym(s):
Potassium Phosphate Monobasic solution, Monopotassium phosphate, Potassium dihydrogen phosphate solution
CAS Number:
Products
Product Category
Brand
Feature
Physical Form
Format
Quality Segment
Application
Reference Material Type
Special Grade
Sterilization Method
Showing 1-2 of 2