
Polyoxyethylene (20) sorbitan monolaurate solution

Synonym(s):
Polyethylene glycol sorbitan monolaurate solution, Polyoxyethylenesorbitan monolaurate
CAS Number:
Products
Product Category
Brand
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing