
Phenyl benzoate

Synonym(s):
Benzoic acid phenyl ester
Linear Formula:
C6H5CO2C6H5
CAS Number:
Molecular Weight:
198.22
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing