
Palladium on carbon

Synonym(s):
Pd/C
Linear Formula:
Pd
Molecular Weight:
106.42
Products
Brand
Sustainability
Feature
Formula Weight
Physical Form
Element
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-12 of 12
Compare
Product Number
Description
Pricing